Cole-Parmer Microcentrifuge with Blue Lid; 2 Rotors
Built-in compartment for storage of PCR rotor and adapters
  • Eight position microtube rotor
  • Storage compartment for PCR rotor
  • Near silent operation
  • Starts and stops in seconds
  • Conserves valuable bench space
This personal-sized microcentrifuge allows for a quick spin down of microtubes and PCR tubes. Rotor changeover is quick and easy—no need for set screws and tools. Features transparent blue lid.
48,700.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max RCF x g Max Speed (rpm) Power (VAC)
Availability Pricing
COMPARE
2000 6000 115
₹48,700.00INR / Each
COMPARE
2000 6000 230
₹48,700.00INR / Each