Cole-Parmer® MBS-200 Series Micro Bead Sterilizers
Removes dangers of open flame
  • Suitable for use in fume hoods because the sterilizers do not use gas, flames, or chemicals
  • A taller model is available for sterilization of larger tools
  • Digital heat control up to 300°C
These micro bead sterilizers are ideal for cleaning of small tools. When filled with glass beads, the temperature can be used to eradicate bacteria, spores, and other microorganisms. An LED display allows you to set temperature precisely with the control knob. Multiple safety features are implemented on this sterilizer. The safety cover can be used to reduce heat loss when ramping which saves energy. The innovative design keeps heat in while the outside remains cool to the touch.
131,000.00 - ₹157,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Chamber Height (in) Power (VAC)
Availability Pricing
COMPARE
3.15 115
₹131,000.00INR / Each
COMPARE
3.15 230
₹131,000.00INR / Each
COMPARE
5.5 115
₹157,000.00INR / Each
COMPARE
5.5 230
₹157,000.00INR / Each