This product is now discontinued, View Related Products
Cole-Parmer Manual Slide Staining Stations
Three-, six-, and twelve-well configurations are available to meet your staining needs
  • Built-in handles allow for convenient moving around the lab
  • Interlocking design allows multiple units to be attached together
Choose the well configuration that meets your application requirements. The metal-extruded frames interlock for back-to-back connections of multiple racks. Stations feature wells that are deep enough to immerse 1" x 3" slides. These plastic wells hold up to 250 mL of reagent, are solvent resistant, and include lids for limiting reagent evaporation. The three-position model has a removable plastic top that covers the entire assembly. Larger sizes feature a see-through, sliding plastic cover that allows access from either end, further reducing the chance of evaporation between staining protocols.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Capacity (Number of slides) External Height (in) External Width (in)
Availability Pricing
COMPARE
3 7 8
Call For Price
COMPARE
6 6 15
Call For Price
COMPARE
12 6 28