Cole-Parmer® LiteSafe Centrifuge Tubes
Protect photosensitive samples
  • Completely opaque - protects even the most light sensitive samples
  • DNase, RNase, and Pyrogen free
  • Ideal for use with fluorescent probes or ethidium bromide
Disposable polypropylene tubes and screw caps have recessed HDPE lids to prevent leaking. Tubes feature a frosted writing surface for permanent identification and labeling. Autoclavable to 122 °C and can be stored down to -100 °C.
4,200.00 - ₹5,100.00INR / Pkg of 50
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max RCF x g Tube Bottom Shape Tube Capacity (mL)
Availability Pricing
COMPARE
8400 Conical 15
₹4,200.00INR / Pkg of 50
COMPARE
10000 Conical 50
₹5,100.00INR / Pkg of 50