This product is now discontinued, View Related Products
Cole-Parmer® Essentials Large-Tip Opening Serological Pipettes
Get fast, accurate delivery
  • Ideal for viscous solutions and tissue culture work
  • Calibrated "to deliver"
  • Permanent amber markings for easy reading
These reusable pipettes are calibrated “to deliver/blow out.” (A “to deliver/blow out” pipette must be allowed to drain, then the drop that remains in the tip must be blown out and added to the original delivery to equal the exact volume.) Top is formed to fit a cotton plug (not included). Pipettes will fit cotton plugging machines. The 1 mL pipette has approximately a 0.1'' (2.5 mm) tip opening; all other sizes have approximately a 0.12'' (3 mm) tip opening. All pipettes are constructed of borosilicate glass, have permanent amber markings, and are specially treated to reduce chipping. Accuracy is printed on each pipette. Sold in cases of 12.
29,800.00 - ₹40,200.00INR / Pkg of 1
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Accuracy (%) Max Volume (mL) Sterile
Availability Pricing
COMPARE
±0.050mL 1 No
Call For Price
COMPARE
±0.025mL 2 No
Call For Price
COMPARE
±0.020mL 5 No
₹29,800.00INR / Pkg of 12
COMPARE
±0.010mL 10 No
Call For Price
COMPARE
±0.200mL 25 No
₹40,200.00INR / Pkg of 12