This product is now discontinued, View Related Products
Cole-Parmer LabGEN 7 Economy Homogenizers
Process tissue samples from 0.25 mL to 30 mL
  • Process most samples in under one minute
  • Handheld or post mountable
  • Easy to clean and disassemble
The economical LabGEN 7 homogenizers are ideal for processing samples ranging from 0.25 to 30 mL. These homogenizers are lightweight to reduce hand strain during extended periods of operation and may be post mounted to an optional stand assembly. The 7-mm generator probe (included) is easy to disassemble and clean. Unit operates with variable-speed motors providing speeds from 5000 to 35,000 rpm and are extremely quiet — under 72 dB.
164,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Max Sample Size (mL) Power (VAC)
Availability Pricing
COMPARE
30 115
₹164,000.00INR / Each
COMPARE
30 220
₹164,000.00INR / Each
COMPARE
30 115
Call For Price
COMPARE
30 220