Cole-Parmer® IBS-200 Series Plus Compact Bench Counting Scale
Basic weighing and simple check weighing in a compact design
  • Large backlit LCD for quick reading
  • Overload protection prevents damage to internal components
  • LED signals is sample is under, within, or over set limits
  • Display hold function freezes display when load is removed
  • Large stainless-steel pan allows for quick cleaning
  • Programmable auto power-off saves energy
These affordable compact bench scales handle basic weighing tasks and check weighing operations with a simple sealed keypad and intuitive operation. They offer five weighing units, percent weighing, simple parts counting, and check weighing. A fully charge battery can operate for up to 240 hours. The scales can also be operated using a USB power source. Other features include external calibration, indicators for stable, zero, net weight, and low battery functions, and zero tracking and tare.
43,200.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Readability (g)
Availability Pricing
COMPARE
3 6.5 0.1
₹43,200.00INR / Each
COMPARE
6 13 0.2
₹43,200.00INR / Each
COMPARE
15 33 0.5
₹43,200.00INR / Each
COMPARE
30 66 1
₹43,200.00INR / Each