Cole-Parmer In-Line or Submersible, High Accuracy pH Electrodes
Unique design provides preductable, reliable, and quick response with changing solution temperatures
  • Ideal for real-time pH measurement and control
  • Electrode drift and accuracy problems are virtually eliminated
  • No premature loss in performance at elevated temperatures
R series electrodes respond to changing temperatures cycle of 25 to 80 °C in as little as 30 seconds, versus 1-3 minutes with conventional electrodes. The unique design and internal fill solution have a low temperature coefficient for rapid response to process changes.
These double junction electrodes are potted in Ryton housing with a semi-dome bulb protector that produces a self-cleaning effect. Electrodes have porous PTFE junctions and your choice of temperature compensation. Operate at temperatures of 0 to 100 °C and pressures up to 100 psi. Housings have a 3/4” NPT(M) threads on both ends for in-line or submersion applications. All electrodes include a 10-ft coaxial cable with BNC connector and tinned ends for models with ATC elements. Insertion depth is 1”.
60,100.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
R-Series pH electrode, 10-ft cable, 100 Ohms Pt RTD
₹60,100.00INR / Each
COMPARE
R-Series pH electrode, 10-ft cable, 100 3K Ohms Balco
₹60,100.00INR / Each