This product is now discontinued, View Related Products
Cole-Parmer Horizontal Mini-Gel Gel Trays
Gel Trays let you pour gels in the unit with the tapered baffles, outside the unit by taping the ends of the tray, or outside the unit using the gel casters sold below. UV-transparent trays are winged for easy removal.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Buffer Solution Required (mL) Max Voltage (VDC) Number of Wells