Cole-Parmer® Essentials Tall-Form Crucible, High Alumina
Use for high temperature work demanding contamination-free results
  • Highly resistant to chemical attack
  • High alumina ware is highly refractory, ideal for use in reducing and oxidizing atmospheres
  • Autoclavable
Unglazed tall-form crucibles are 99.7% pure aluminum oxide. Withstand temperatures up to 1750 °C (3182 °F). High alumina labware temperature change rate should not exceed 150 °C/hour.
5,000.00 - ₹13,600.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Top Outside Diameter (mm)
Availability Pricing
COMPARE
10 30
₹5,000.00INR / Each
COMPARE
20 39
₹6,600.00INR / Each
COMPARE
50 52
₹8,600.00INR / Each
COMPARE
100 65
₹13,600.00INR / Each