This product is now discontinued, View Related Products
Cole-Parmer Heating Controllers
Compact and cost effective
  • Ideal for use on benchtop or mount to frame
One-way external controllers provide manual control of heating mantles, electric Bunsens, and heating tapes and cords. The controllers are designed to regulate the heating of resistive loads for one piece of laboratory equipment at a time. A regulator control dial increases power to the equipment as it is turned clockwise. When the power is on, a white neon indicator lights up. As power is being supplied to the equipment an amber neon indicator begins to pulsate. Once maximum power is reached, the amber neon becomes fully illuminated.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Analog Heating Controller, 450 W, 120 VAC
Call For Price
COMPARE
Analog Heating Controller, 800 W, 230 VAC
Call For Price
COMPARE
Analog Heating Controller, 1150 W, 120 VAC
Call For Price
COMPARE
Analog Heating Controller, 1800 W, 230 VAC