Cole-Parmer Half-Cell pH and ORP Electrodes
Combine the half-cell and reference electrodes to create a system tailored to meet your specific application requirements
  • Refillable reference electrodes allow for repeated uses
These half-cell electrodes have BNC connectors and must be used with reference electrodes, available as 05990-60 or 05899-25. Both of these electrodes are made of blue sensing glass, have a 212°F (100°C) maximum temperature and are 154 mm long by 12 mm in diameter. Model 05990-15 is for general purpose pH use and 05990-16 is for general purpose ORP measurment.
13,600.00 - ₹22,400.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Electrode Type
Availability Pricing
COMPARE
Half-cell; single junction
₹13,600.00INR / Each
COMPARE
Single junction
₹22,400.00INR / Each