Cole-Parmer® Grounded pH Electrodes
Ground loop interrupt eliminates electrical interference
  • Fluid flow across the surface provides a cleaning action that extends the life of the electrode and improves performance
  • Flat measuring surface minimizes electrode coating, scratching, and breakage
This submersible electrode is ideal when your system calibrates outside your tank but gives incorrect readings when installed. Combat interference with a solution ground and internal battery powered ground loop interrupt. Built-in amplifier in electrode cartridge permits long cable runs.
113,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
ATC Connector Connector Electrode Body Material
Availability Pricing
COMPARE
BNC CPVC
₹113,000.00INR / Each