Cole-Parmer® Graduated Glass Cylinder, Class B
Accurate measurements even at lowest graduations
  • Calibrated “To Contain”
  • Single metric scale
Cylinders provide accurate measurements even at the lowest graduations. These cylinders are used in ASTM tests D 86 and D447. Models 34200-04 and -05 meet ASTM E 133 requirements. Cylinders are made of borosilicate glass and feature easy-to-read white ceramic scales and handy pouring spouts. Heavy, round bases on the smaller sizes and a hexagonal base on the 2000 mL size ensure stability on your benchtop.
860.00 - ₹11,800.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
COMPARE
10 0.2 Glass
₹1,400.00INR / Pkg of 2
COMPARE
25 0.5 Glass
₹860.00INR / Each
COMPARE
50 1 Glass
₹980.00INR / Each
COMPARE
100 1 Glass
₹1,200.00INR / Each
COMPARE
250 2 Glass
₹2,200.00INR / Each
COMPARE
500 5 Glass
₹3,600.00INR / Each
COMPARE
1000 10 Glass
₹5,000.00INR / Each
COMPARE
2000 20 Glass
₹11,800.00INR / Each
Showing 1- 8 of 8