Cole-Parmer® Glass Graduated Cylinders
Hexagonal base prevents cylinder from tipping over
  • Durable Borosilicate 3.3 Glass
  • Easy read graduations
  • Withstands wide temperature range -50 to 550ºC
  • Graduated tolerances from ±0.1 to ±20 depending on size
  • Autoclavable
800.00 - ₹6,300.00INR / Pkg of 1
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Tolerance (mL)
Availability Pricing
COMPARE
5 0.1 ±0.1
₹1,600.00INR / Pkg of 2
COMPARE
10 0.2 ±0.2
₹1,300.00INR / Pkg of 2
COMPARE
25 0.5 ±0.5
₹800.00INR / Pkg of 1
COMPARE
50 1 ±1
₹800.00INR / Pkg of 1
COMPARE
100 1 ±1
₹1,100.00INR / Pkg of 1
COMPARE
250 2 ±2
₹1,700.00INR / Pkg of 1
COMPARE
500 5 ±5
₹2,700.00INR / Pkg of 1
COMPARE
1000 10 ±10
₹3,600.00INR / Pkg of 1
COMPARE
2000 20 ±20
₹6,300.00INR / Pkg of 1
Showing 1- 9 of 9