This product is now discontinued, View Related Products
Cole-Parmer Glass Fermentation Dissolved Oxygen Probes
Low-cost, glass-body design
  • Polarographic
These economical glass-body probes are ideal for your laboratory fermentation applications where sterilization takes place in a separate autoclave. Probes feature a threaded detachable cable lead. Order appropriate cable separately.
32,700.00 - ₹36,600.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Fermentation DO Probe; Glass/Polarographic/180 mm
₹32,700.00INR / Each
COMPARE
Fermentation DO Probe; Glass/Polarographic/210 mm
Call For Price
COMPARE
Fermentation DO Probe; Glass/Polarographic/300 mm
₹36,600.00INR / Each