This product is now discontinued, View Related Products
Cole-Parmer Full-Size, Dual-Power Stereomicroscopes
Larger stage accommodates bigger specimens
These full-size, dual-power stereomicroscopes stand 13'' tall and have a working distance of 84 mm. They feature a 45° inclined swivel head that rotates 360° and has adjustable diopter and interpupillary controls. Paired objectives are mounted in a revolving turret. Change magnification by rotating the turret 90°. Paired 10x widefield eyepieces have a 34-mm OD and accept reticles with 22.8-mm OD. The specially designed lamp directs light exactly where you want. The incident (top) light source has an adjustable beam and variable intensity control. The transmitted (bottom) light source is fluorescent giving you optimum light and cool temperatures to prolong the life of a live specimen. Microscopes are supported by a cast metal frame with gray enamel finish. Each stereomicroscope is supplied with both frosted glass and reversible black-and-white stage plates and a vinyl dust cover.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Magnification Power (VAC)
Availability Pricing
COMPARE
10x/30x 115
Call For Price
COMPARE
10x/30x 230
Call For Price
COMPARE
20x/40x 115
Call For Price
COMPARE
20x/40x 230