This product is now discontinued, View Related Products
Cole-Parmer Filter Pipette Tips 72206
Cleanroom produced for ready-to-use applications
  • Slim, thin-walled tips feature graduations
  • Filter protects samples and pipettor against contamination
  • Certified RNase, DNase, and endotoxin free
  • Autoclavable at 121°C for 20 minutes
Constructed of pure, high-quality polypropylene (PP) without additives, these tips are designed to give high levels of accuracy, precision, and reproducibility. The smooth surface minimizes binding of enzymes and nucleic acids to the tip wall.
The non-self-sealing filter plug prevents aerosols and liquids from being aspirated into the body of the pipettor. Filter is made of pure polyethylene (PE), free from chemical additives, and will not swell.
Tips are available in a storage box with 96-tip rack insert and hinged lid. Order environmentally friendly refill racks separately.
Call For Price
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
Compareitem specifications
Max Volume (µL) Min Volume (µL) Sterile
Availability Pricing
COMPARE
1 0.1 No
Call For Price
COMPARE
10 0.5 No
Call For Price
COMPARE
20 1 No
Call For Price
COMPARE
20 2 No
Call For Price
COMPARE
100 5 No
Call For Price
COMPARE
200 5 No
Call For Price
COMPARE
1000 50 No
Call For Price
COMPARE
1 0.1 Yes
Call For Price
COMPARE
10 0.5 Yes
Call For Price
COMPARE
20 1 Yes
Call For Price
COMPARE
20 2 Yes
Call For Price
COMPARE
100 5 Yes
Call For Price
COMPARE
200 5 Yes
Call For Price
COMPARE
1000 50 Yes
Call For Price
Showing 1- 14 of 14