Cole-Parmer® Essentials Cryogenic Vial with External Thread, Sterile
Stores samples as low as -196°C
  • DNase, RNase, and pyrogen free
  • Radiation sterilized
These polypropylene cryogenic vials feature a self-standing star base designed for easy one-handed manipulation when used with a support rack. Vials include an internal U-shaped bottom, large white frosted labeling area, silk screen graduations, and a leak-proof HDPE cap.
Ideal for used in vapor phase of liquid nitrogen but are not recommended for use in the liquid phase of liquid nitrogen.
33,100.00 - ₹39,600.00INR / Case of 500
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (mm) Tube Bottom Shape Volume (mL)
Availability Pricing
COMPARE
48.5 Star base 1.8
₹39,600.00INR / Case of 1000
COMPARE
91.5 Star base 4.5
₹33,100.00INR / Case of 500