Cole-Parmer® Essentials Double Bagged Sterile Lab Sampling Scoops, Polystyrene
Double bagged for extra protection from contamination
  • Specifically designed for multi-zone clean rooms
  • Gamma irradiation sterilized (SAL 10-6)
  • Meets FDA guidelines for food, drug, and cosmetic sampling
  • Individually sealed easy-to-open bags all packaged within an outer bag
These single-use, sterile lab scoops help reduce cross contamination and can be used to hold a variety of media. Designed for use in multi-zone cleanrooms, they are individually bagged then packaged within another bag for extra contamination protection. They are manufactured from sturdy, one-piece, FDA- grade polystyrene plastic, and have an ergonomic, comfortably curved handle to make sample retrieval easy. A flat bottom allows the sampling scoop to be placed on a surface without spillage and to be used as a weighing dish.
These laboratory scoops are gamma irradiation sterilized (SAL 10-6); free of BSE/TSE, heavy metals, BPA, and phthalates; and lot traced. A Certificate of Irradiation is available upon request. Specified volumes are approximate and not intended for measuring specific volumes.
8,800.00 - ₹12,100.00INR / Case of 50
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Overall Length (cm) Width (cm)
Availability Pricing
COMPARE
60 20.21 3.92
₹10,500.00INR / Case of 50
COMPARE
120 22.01 4.91
₹12,100.00INR / Case of 50
COMPARE
250 24.66 7.83
₹8,800.00INR / Case of 25