Cole-Parmer® Essentials Sterile Media Bottles
Reduce risk of cross contamination with disposable Cole-Parmer media bottles
  • Shatterproof PET provides a safe alternative to glass
  • Pliable material ensures comfortable grip
  • Supplied with HDPE screw caps—free of heavy metals
  • Individually packed and EB sterilized
  • Certificate of Conformance available upon request
Bottle is embossed with graduations on both the sides for easy reference and carries 2-years shelf life from the month of production. Bottles can used sustain temperature range from -60°C to 60°C.
14,400.00 - ₹17,700.00INR / Case of 100
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Sterile PET Media Bottle, 250 mL; 100/Cs
₹14,600.00INR / Case of 100
COMPARE
Sterile PET Media Bottle, 500 mL; 100/Cs
₹17,700.00INR / Case of 100
COMPARE
Sterile PET Media Bottle, 1000 mL; 48/Cs
₹14,400.00INR / Case of 48