Cole-Parmer® Essentials Double-Ended Cell Lifters, Sterile, HDPE
Double-ended cell lifter design provides two tools in one for quick removal of cell layers
  • Easily harvest cells from culture dishes, plates, and tubes
  • 19 mm beveled edge flat blade ensures even contact with growth surface
  • Sterilized by gamma irradiation
  • Individually packaged
Double-ended lifters allow easy harvest of cells from culture dishes, plates and tubes. Both versions have a 19 mm beveled edge flat blade on one end. Second end choices include a narrow-tapered blade for 96-well plates or microcentrifuge tubes, or a J-hook to vertically lift cells from 4- to 48-well plates. Made of high-density polyethylene, these cell lifters are individually wrapped and sterilized by gamma irradiation, and are RNase/DNase free, nonpyrogenic, and noncytotoxic.
15,700.00INR / Case of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Overall Length (in) Sterile Description
Availability Pricing
COMPARE
9 Yes Cell Lifters, Sterile, HDPE, Flat and Narrow Blades; 100/Cs
₹15,700.00INR / Case of 100
COMPARE
9 Yes Cell Lifters, Sterile, HDPE, Flat Blade and J-hook; 100/Cs
₹15,700.00INR / Case of 100