Cole-Parmer® Essentials Spray Bottles
Make awkward wash bottles obsolete
  • Provide controlled delivery—comfortable for one-handed operation
  • Ideal for hot, cold, aqueous, and solvent-based solutions
  • Contoured to fit in the hand
Set nozzle to fine mist for TLC plates, standard spray for rinsing glassware, or continuous stream for a forceful jet when washing filters. Nozzle also has closed position to prevent contamination. Bottles are constructed of translucent HDPE and come with PP cap and spray assembly.
2,500.00 - ₹4,100.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Autoclavable Capacity (mL) Cap Color
Availability Pricing
COMPARE
No 1000 Red and white
₹4,100.00INR / Each
COMPARE
No 240 Blue and white
₹2,500.00INR / Pkg of 3