Cole-Parmer® Essentials Sterile Oval Lab Sampling Spoons, Polystyrene
High-quality, chemically resistant polystyrene
  • Gamma irradiation sterilized
  • DNase, RNase, and pyrogen-free
  • Withstands temperatures between -10 to +70 °C (14 to 158 °F)
These high-quality, oval shaped, polystyrene lab spoons are resistant to most acids, bases, and commonly used solvents. The easy-to-grip long handles allow efficient sampling of various powders, granulates, and pastes while reducing the risk of cross contamination. The handles are also slightly inclined to allow the sampling spoons to lie horizontally on a flat surface.
Individually packaged, printed with lot number, production date, and expiration date, these gamma- irradiated, sterile spoons are DNase, RNase, and pyrogen-free. They are guaranteed to withstand temperatures between -10 to 70 °C. (14 to 158 °F) and are intended for one-time use and disposal. Specified volumes are printed on the handles but are approximate and not intended for measuring specific volumes.
8,600.00 - ₹9,900.00INR / Case of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Overall Length (cm) Width (cm)
Availability Pricing
COMPARE
2.5 12.5 2.9
₹8,600.00INR / Case of 100
COMPARE
10 17.1 4
₹9,900.00INR / Case of 100