Cole-Parmer® Essentials Pre-Cleaned Wide-Mouth Plastic Bottle, HDPE
Leakproof design keeps samples safe
  • Pre-cleaned according to EPA standards
These wide-mouth high-density polyethylene (HDPE) bottles are leak-proof and break-resistant — excellent for soil, sludge, and sediment samples. Bottles are pre-cleaned according to Environmental Protection Agency (EPA) standards and certified to meet the EPA’s Performance Based Specifications for metals analysis and water quality parameters.
19,300.00 - ₹31,700.00INR / Case of 24
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
COMPARE
125 38-415 High Density Polyethyene (HDPE)
₹25,200.00INR / Case of 72
COMPARE
250 43-415 High Density Polyethyene (HDPE)
₹31,700.00INR / Case of 72
COMPARE
500 53-415 High Density Polyethyene (HDPE)
₹27,600.00INR / Case of 48
COMPARE
1000 63-415 High Density Polyethyene (HDPE)
₹19,300.00INR / Case of 24