Cole-Parmer® Essentials Powder Sample Collector, Sterile HDPE
Quickly and easily take a sample at a specified point within the vessel
  • Manufactured in Class 100,000 cleanroom and individually packaged
  • Constructed of chemically inert high-density polyethylene (HDPE)
These PowderThief powder samplers are designed to take representative samples from known locations within the bulk powder. The samplers are manufactured, assembled, and packed in a Class 100,000 Cleanroom to ensure cleanliness and individually bagged. Samplers are also sterilized via gamma irradiation (minimum dose 25 kGy).
50,500.00 - ₹66,400.00INR / Pkg of 20
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (cm) Sterile Volume (mL)
Availability Pricing
COMPARE
50 Yes 10
₹50,500.00INR / Pkg of 20
COMPARE
100 Yes 10
₹66,400.00INR / Pkg of 20
COMPARE
50 Yes 20
₹50,500.00INR / Pkg of 20
COMPARE
100 Yes 20
₹66,400.00INR / Pkg of 20