Cole-Parmer® Essentials Polypropylene 15 and 50 mL Vial Storage Boxes
Accommodate 15- or 50-mL centrifuge tubes for long-term storage
  • Store down to -90°C (-130°F) or autoclave up to 122°C (252°F)—wide temperature range
  • Inspect, reference, and identify samples through the and clear lid and easy-to-read numeric grid
  • Label or mark boxes on the white top portion
  • Compatible with most 4" stainless steel freezer racks
These cryogenic storage boxes offer excellent low-temperature storage. Polypropylene boxes have clear lids with colored inserts and numbered grids to facilitate easy sample identification.
3,300.00 - ₹3,600.00INR / Pkg of 2
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubes Held Tube Size Description
Availability Pricing
COMPARE
36 15-mL Polypropylene Freezer Box, 36 x 15 mL, Blue; 2/Pk
₹3,600.00INR / Pkg of 2
COMPARE
16 50-mL Polypropylene Freezer Box, 16 x 50 mL, Orange; 2/Pk
₹3,300.00INR / Pkg of 2