Cole-Parmer® Sterile Media Bottles, Square Plastic, PET
Sterile, plastic media storage bottles ideal for safe storage and transportation of cell culture media
  • Crush-resistant corners
  • Embossed volume graduations
  • Space saving square shape
  • Neck shape allows easy filling and pouring
Recommended for safe storage and transportation of cell culture media, these sterile, plastic bottles feature thick, transparent walls and crush-resistant corners. Sterilized by gamma irradiation, they are ideal for use when trying to prevent sample contamination. They feature easy to see, embossed graduations on two sides, and a neck shape that allows for easy filling and pouring. The space saving, square shape helps you maximize shelf space.
Made of polyethylene terephthalate (PET), they have embossed volume graduations for simple liquid level viewing. The polyethylene caps have large ridges for fast closure.
32,700.00 - ₹42,500.00INR / Case of 96
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Overall Height (cm)
Availability Pricing
COMPARE
125 25 10.5
₹39,500.00INR / Case of 144
COMPARE
250 25 14.7
₹32,700.00INR / Case of 96
COMPARE
500 50 17.4
₹42,500.00INR / Case of 96
COMPARE
1000 50 21.4
₹39,300.00INR / Case of 48