Cole-Parmer® Essentials Easy Flip™ Centrifuge Tubes
No more lost caps!
  • Made from high grade gamma resistant polypropylene
  • Maximum RCF: 9,400xg
  • Chloroform resistant black printed graduations and white writing area for unique identification
  • Autoclavable at 121°C and freezable to -80°C
  • Leak proof design to avoid cross contamination
  • Gamma sterilized
  • Racked tubes are packed in environmental friendly recyclable chipboard racks
  • Bagged tubes are packed in durable and re-sealable (zip closure) bags
  • Inner packaging is individually labelled for lot traceability
11,400.00 - ₹25,900.00INR / Case of 1
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem Availability Pricing
COMPARE
₹11,500.00INR / Case of 1
COMPARE
₹11,400.00INR / Case of 1
COMPARE
₹17,500.00INR / Case of 1
COMPARE
₹19,900.00INR / Case of 1
COMPARE
₹15,700.00INR / Case of 1
COMPARE
₹15,500.00INR / Case of 1
COMPARE
₹23,100.00INR / Case of 1
COMPARE
₹25,900.00INR / Case of 1
Showing 1- 8 of 8