Cole-Parmer® Essentials Double-Ended Lab Sampling Spoons, 304 Stainless Steel
One utensil; two capacity use
  • Chemically resistant 304 stainless steel
  • Smooth polished and easy to clean
  • BSE/TSE free
Double ended lab sampling spoons include a smaller capacity spoon at one end, and larger capacity spoon at the other. This allows you to take different size samples without changing utensil; simply flip the utensil around. Ideal for sampling powder and liquid, these BSE/TSE free, 304 stainless steel lab sampling spoons have excellent chemical compatibility. They are polished smooth to ensure easy and thorough cleaning and can be autoclaved for sterility if needed. When used in place of disposable spoons, these reusable lab spoons help reduce waste in the laboratory, as well as save you money and reordering time.
2,100.00 - ₹25,000.00INR / Set of 3
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Overall Length (cm) Description
Availability Pricing
COMPARE
8 30 Double-Ended Lab Sampling Spoons, 304 Stainless Steel, Set of 3 (0.5 to 8 mL)
₹25,000.00INR / Set of 3
COMPARE
2 15 Double-Ended Lab Sampling Spoons, 304 Stainless Steel, 0.5 mL and 2 mL
₹2,100.00INR / Each
COMPARE
2.8 18 Double-Ended Lab Sampling Spoons, 304 Stainless Steel, 0.8 mL and 2.8 mL
₹2,200.00INR / Each
COMPARE
3 21 Double-Ended Lab Sampling Spoons, 304 Stainless Steel, 1.6 mL and 3 mL
₹2,700.00INR / Each
COMPARE
5 25 Double-Ended Lab Sampling Spoons, 304 Stainless Steel, 1.8 mL and 5 mL
₹3,000.00INR / Each
COMPARE
8 30 Double-Ended Lab Sampling Spoons, 304 Stainless Steel, 2 mL and 8 mL
₹3,500.00INR / Each