Cole-Parmer® Essentials Cleaning Brushes
Long handle brushes allow for a thorough cleaning
  • Brush heads conveniently screw on to the handle
  • Constructed with a sturdy aluminum handle and nylon bristles
These cleaning brushes are ideal for cleaning both stainless steel, plastic, and glass samplers. Brushes heads are made with nylon Bristles in a twisted wire frame and are 100 mm long. The durable aluminum handles come in 500 mm lengths and screw together for a total handle length of 1000 mm.
Two brush head sizes are available. The 35 mm diameter brush is suitable for cleaning a vessel with a 15 to 30 mm diameter. The 50 mm diameter brush is suitable for cleaning a vessel with a 30 to 45 mm diameter.
4,100.00 - ₹4,700.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (in) Length (in) Description
Availability Pricing
COMPARE
1.38 43 5/16 Cleaning Brush, 1100 mm Length x 35 mm Diameter
₹4,100.00INR / Each
COMPARE
1.97 43 5/16 Cleaning Brush, 1100 mm Length x 50 mm Diameter
₹4,700.00INR / Each