Cole-Parmer® Essentials 100-Place Polypropylene Storage Boxes
Featuring reinforced lock and seal for secure storage
  • Reinforced triple hinges prevent cracks or breaks
  • Bold alpha-numeric printing allows accurate tracking of samples
  • Click fit lock ensures box will not open accidentally—large clasp allows easy opening with gloves
Boxes withstand temperatures from -80°C to 121°C, and will not degrade after repeated thaws. Each box features an alphanumeric grid on the exterior and interior, stacking rings and a matte surface suitable for markers or labels. Available with clear or fluorescent colored lids with clear bottoms for easy visual identification. Boxes accommodate 0.5, 1.5, and 2.0 mL microtubes and most brands of cryogenic vials. Black box is suitable for light sensitive samples.
6,100.00 - ₹64,400.00INR / Pkg of 5
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Tube Size Description
Availability Pricing
COMPARE
Black 0.5, 1.5, and 2 mL 100-Place PP Mictrotube Storage Box, Black; Pack of 5
₹6,400.00INR / Pkg of 5
COMPARE
Natural 0.5, 1.5, and 2 mL 100-Place PP Mictrotube Storage Box, Natural; Pack of 5
₹6,100.00INR / Pkg of 5
COMPARE
Blue 0.5, 1.5, and 2 mL 100-Place PP Mictrotube Storage Box, Blue; Pack of 5
₹6,400.00INR / Pkg of 5
COMPARE
Green 0.5, 1.5, and 2 mL 100-Place PP Mictrotube Storage Box, Green; Pack of 5
₹6,400.00INR / Pkg of 5
COMPARE
Orange 0.5, 1.5, and 2 mL 100-Place PP Mictrotube Storage Box, Orange; Pack of 5
₹6,400.00INR / Pkg of 5
COMPARE
Purple 0.5, 1.5, and 2 mL 100-Place PP Mictrotube Storage Box, Purple; Pack of 5
₹6,400.00INR / Pkg of 5
COMPARE
Assorted 0.5, 1.5, and 2 mL 100-Place PP Mictrotube Storage Box, Assorted; Pack of 5
₹6,400.00INR / Pkg of 5
COMPARE
Natural 0.5, 1.5, and 2 mL 100-Place PP Mictrotube Storage Box, Natural; Pack of 5
₹60,700.00INR / Case of 50
COMPARE
Blue 0.5, 1.5, and 2 mL 100-Place PP Mictrotube Storage Box, Blue; Pack of 5
₹60,700.00INR / Case of 50
COMPARE
Green 0.5, 1.5, and 2 mL 100-Place PP Mictrotube Storage Box, Green; Pack of 5
₹60,700.00INR / Case of 50
COMPARE
Orange 0.5, 1.5, and 2 mL 100-Place PP Mictrotube Storage Box, Orange; Pack of 5
₹64,400.00INR / Case of 50
COMPARE
Purple 0.5, 1.5, and 2 mL 100-Place PP Mictrotube Storage Box, Purple; Pack of 5
₹60,700.00INR / Case of 50
COMPARE
Black 0.5, 1.5, and 2 mL 100-Place PP Mictrotube Storage Box, Black; Case of 50
₹60,700.00INR / Case of 50
Showing 1- 13 of 13