Cole-Parmer Epoxy-Body pH Electrodes for Field Use
Ideal for your most rugged applications
  • Ceramic reference-junction pH electrodes (clog resistant)
These combination electrodes withstand the most demanding field conditions—the semi-domed tip protects the pH bulb and ensures a quick response when taking measurements in harsh and dirty environments. Easy-to-clean electrodes feature an impact-resistant epoxy body for further protection from the elements. The large, annular ceramic reference junction and sealed cross-linked polymer reference resist clogging.
Operating range is from 0 to 14 pH. Electrodes measure 110 mm L x 12 mm dia and withstand temperatures up to 212°F (100°C). All models have BNC connectors.
16,900.00 - ₹21,900.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
pH Electrodes for Field Use; single-junction 3 foot cable
₹16,900.00INR / Each
COMPARE
pH Electrodes for Field Use; double-junction 3 foot cable
₹20,300.00INR / Each
COMPARE
pH Electrodes for Field Use; single-junction 10 foot cable
₹18,600.00INR / Each
COMPARE
pH Electrodes for Field Use; double-junction 10 foot cable
₹21,900.00INR / Each