Cole-Parmer® Essentials Class A Glass Volumetric Flasks
Tight tolerance ensures accuracy
  • White easy read marking for volume measurement
  • Volumetric flasks feature right tolerances from ±0.15 to ±1.2 depending on size
  • Meets the ASTM E 438 Type I Class A standard
  • Autoclavable
These Cole-Parmer® Essentials volumetric flasks are constructed of quality borosilicate 3.3 glass. These flasks are calibrated "To Contain" (IN) by DIN standard at 20°C. Flasks can withstand a wide temperature range from –50 to 550°C.
1,300.00 - ₹26,500.00INR / Pkg of 2
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Tolerance (mL) Description
Availability Pricing
COMPARE
2 ±0.15 Volumetric Flask, Glass, with Glass Stopper, 2 mL, 10/pk
₹5,800.00INR / Pkg of 10
COMPARE
5 ±0.02 Volumetric Flask, Glass, with Glass Stopper, 5 mL, 10/pk
₹5,100.00INR / Pkg of 10
COMPARE
10 ±0.025 Volumetric Flask, Glass, with Glass Stopper, 10 mL, 2/pk
₹3,000.00INR / Pkg of 2
COMPARE
20 ±0.030 Volumetric Flask, Glass, with Glass Stopper, 20 mL, 2/pk
₹1,400.00INR / Pkg of 2
COMPARE
25 ±0.04 Volumetric Flask, Glass, with Glass Stopper, 25 mL, 2/pk
₹1,500.00INR / Pkg of 2
COMPARE
50 ±0.05 Volumetric Flask, Glass, with Glass Stopper, 50 mL, 2/pk
₹1,300.00INR / Pkg of 2
COMPARE
100 ±0.1 Volumetric Flask, Glass, with Glass Stopper, 100 mL, 2/pk
₹1,900.00INR / Pkg of 2
COMPARE
200 ±0.15 Volumetric Flask, Glass, with Plastic Stopper, 200 mL, 2/pk
₹2,200.00INR / Pkg of 2
COMPARE
250 ±0.15 Volumetric Flask, Glass, with Glass Stopper, 250 mL, 2/pk
₹2,700.00INR / Pkg of 2
COMPARE
500 ±0.2 Volumetric Flask, Glass, with Glass Stopper, 500 mL, 2/pk
₹3,800.00INR / Pkg of 2
COMPARE
1000 ±0.4 Volumetric Flask, Glass, with Glass Stopper, 1000 mL, 1/pk
₹3,800.00INR / Pkg of 1
COMPARE
2000 ±0.6 Volumetric Flask, Glass, with Glass Stopper, 2000 mL, 1/pk
₹10,600.00INR / Pkg of 1
COMPARE
5000 ±1.2 Volumetric Flask, Glass, with Glass Stopper, 5000 mL, 1/pk
₹26,500.00INR / Pkg of 1
Showing 1- 13 of 13