This product is now discontinued, View Related Products
Cole-Parmer Electrophoretic Blotting Systems 16108
For Western, Southern, or Northern blots – transfer proteins and nucleic acids from agarose or acrylamide gels
Glass cooling chamber with tubing adapters is built into base for temperature-controlled runs. Platinum wire electrodes are mounted in recessed grooves for protection from damage when cleaning unit or loading cassettes. Electrode design provides even current distribution over entire gel surface. Place a magnetic stirring bar beneath cassettes to circulate buffer within unit for more efficient cooling. Order additional cassettes separately.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Buffer Solution Required (mL) Description
Availability Pricing
COMPARE
3800 Electrophoretic Blotting System, 3 Cassette capacity
Call For Price
COMPARE
1100 Electrophoretic Blotting System, 4 Cassette capacity