Cole-Parmer® Electronic Bottletop Burettes
Get better accuracy with motor-controlled filling and dispensing
  • Easy, user-friendly, TFT touch screen helps to perform quick and accurate titrations
  • Motor-controlled piston movement, help in eliminating the errors
  • Three pre-set dispensing speeds allows achieve the end point very accurately.
  • Conforms to ISO 8655 standards & GLP compliant.
Accuracy of Cole-Parmer E-Burette is ± 0.2% and able to perform precise titrations within the error limits of 'Class A' glass burettes. 20 results can be saved in the memory. Computer connectivity is available through a micro-USB cable for transferring and recording data. Rechargeable, long-life, replaceable batteries are provided in the control panel. Amber colored window is provided as an accessory for those chemicals/reagents that are sensitive to light
239,000.00 - ₹243,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Power (VAC) Description
Availability Pricing
COMPARE
10 230 Electronic Burette, Bottletop Dispenser, 10 mL, 230V, 50Hz
₹239,000.00INR / Each
COMPARE
25 230 Electronic Burette, Bottletop Dispenser, 25 mL, 230V, 50Hz
₹243,000.00INR / Each
COMPARE
50 230 Electronic Burette, Bottletop Dispenser, 50 mL, 230V, 50Hz
₹243,000.00INR / Each