Cole-Parmer® Electrode Holders for 12-mm Glass Body Electrode with Integrated Threaded Assemblies
Easily remove electrodes in seconds without removing the holder
  • Ideal for in-line or submersion monitoring
Integral cap assembly permits quick servicing for calibrations or replacement. Electrode holders have threads on each end. Install holders into a pipe tee for in-line use or on the end of a submersion pipe. Choose from CPVC for economical aplications or PVDF for enhanced chemical resistance. Electrodes order separately.
9,100.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Electrode Body Material
Availability Pricing
COMPARE
CPVC
₹9,100.00INR / Each