This product is now discontinued, View Related Products
Cole-Parmer Dual-Line Process Meters and Rate/Totalizers
Get twice as much information for better evaluation and control
  • Large dual-line 6-digit display is easy to read
  • Three easy programming options—front-panel buttons, copy function, or a PC
  • Front panel is NEMA 4X, IP65 rated
These 1⁄8-DIN panel meters accept 0 to 20 mA, 4 to 20 mA, 0 to 5 V, 1 to 5 V, and ±10 V inputs. They calculate math functions for flow and round horizontal tanks; in addition to
32-point, square root, or exponential linearization. Units can also control several pumps and
alternate controllers. Programming and setup can be easily performed with either the front-panel push buttons, the included MasterView® software, or by using the copy function.
81,800.00 - ₹124,000.00INR / Each
50 variations of this product are available.
See All Product Options Below
50 PRODUCT OPTIONS
Compareitem specifications
Control Input VDC DIN Size Description
Availability Pricing
COMPARE
0-5, 1-5 (selectable) 1/8\" Process meter, no relays, 85 to 265 VAC power
₹81,800.00INR / Each
COMPARE
0-5, 1-5 (selectable) 1/8\" Process meter, two SPDT relays, 85 to 265 VAC power
₹92,700.00INR / Each
COMPARE
0-5, 1-5 (selectable) 1/8\" Process meter, 4 to 20 mA output, 85 to 265 power
₹97,100.00INR / Each
COMPARE
0-5, 1-5 (selectable) 1/8\" Process meter, four SPDT relays, 85 to 265 VAC power
₹102,000.00INR / Each
COMPARE
0-5, 1-5 (selectable) 1/8\" Process meter, two SPDT relays and 4 to 20 mA output, 85 to 265 VAC power
₹106,000.00INR / Each
COMPARE
0-5, 1-5 (selectable) 1/8\" Process meter, four SPDT relays and 4 to 20 mA output, 85 to 265 VAC power
₹114,000.00INR / Each
COMPARE
0-5, 1-5 (selectable) 1/8\" Process meter, no relays, 12 to 24 VDC power
₹91,400.00INR / Each
COMPARE
0-5, 1-5 (selectable) 1/8\" Process meter, two SPDT relays, 12 to 24 VDC power
Call For Price
COMPARE
0-5, 1-5 (selectable) 1/8\" Process meter, 4 to 20 mA output, 12 to 24 VDC power
Call For Price
COMPARE
0-5, 1-5 (selectable) 1/8\" Process meter, four SPDT relays, 12 to 24 VDC power
Call For Price
COMPARE
0-5, 1-5 (selectable) 1/8\" Process meter, two SPDT relays and 4 to 20 mA output, 12 to 24 VDC power
₹115,000.00INR / Each
COMPARE
0-5, 1-5 (selectable) 1/8\" Process meter, four SPDT relays and 4 to 20 mA output, 12 to 24 VDC power
₹124,000.00INR / Each
COMPARE
0-5, 1-5 (selectable) 1/8\" Temperature meter, no relays, 85 to 265 VAC power
₹81,800.00INR / Each
COMPARE
0-5, 1-5 (selectable) 1/8\" Temperature meter, two SPDT relays, 85 to 265 VAC power
Call For Price
COMPARE
0-5, 1-5 (selectable) 1/8\" Temperature meter, 4 to 20 mA output, 85 to 265 VAC power
Call For Price
COMPARE
0-5, 1-5 (selectable) 1/8\" Temperature meter, four SPDT relays, 85 to 265 VAC power
₹102,000.00INR / Each
COMPARE
0-5, 1-5 (selectable) 1/8\" Temperature meter, two SPDT relays and 4 to 20 mA output, 85 to 265 VAC power
₹106,000.00INR / Each
COMPARE
0-5, 1-5 (selectable) 1/8\" Temperature meter, four SPDT relays and 4 to 20 mA output, 85 to 265 VAC power
₹114,000.00INR / Each
COMPARE
0-5, 1-5 (selectable) 1/8\" Temperature meter, no relays, 12 to 24 VDC power
Call For Price
COMPARE
0-5, 1-5 (selectable) 1/8\" Temperature meter, two SPDT relays, 12 to 24 VDC power
Call For Price
Showing 1 - 20 of 50