Cole-Parmer® Double-Junction pH and ORP Electrodes with Removable Electrode Assemblies
Easily remove electrodes in seconds without removing the holder
  • Ideal for in-line or submersion monitoring
Double-junction electrodes prevent electrode contamination by solutions that might react with Ag+ or Cl- (heavy metals, organics, and sulfides). Use for measurements up to 13 pH. Withstands temperatures form 23 to 212ºF (-5 to 100ºC).
27,300.00 - ₹36,200.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Removable Blue Glass Double-Junction pH Electrode
₹27,300.00INR / Each
COMPARE
Removable Blue Glass Double-Junction pH Electrode, 100 Ohms RTD
₹36,000.00INR / Each
COMPARE
Integrated Assembly pH Probe, DJ/100Ohm RTD; Plug/Tinned
₹36,200.00INR / Each