Cole-Parmer® Essentials Lab Inoculation Needles, Sterile, Polystyrene
Sterile lab inoculating needles save time and eliminate cross-contamination when transferring microbial organisms
  • Use straight from the pack without sterilizing
  • Flexible and disposable
  • Treated, hydrophilic surface for easier attachment of cells
Ideal for picking single colonies, these disposable polystyrene lab inoculating needles are vacuum-gas plasma surface treated to allow culture medium to attach. The long, smooth, flexible stem can be bent allowing access to narrow or small-volume tubes and dishes.
Manufactured under ISO 13485 and ISO 9001 quality management systems in an ISO Class 8 (formerly Class 100,000) cleanroom environment, these radiation sterilized (SAL 10-6) inoculating needles are available in individual peel-to-open packs, or in resealable packs of 10 or 25. Every package is printed with a lot number for quality traceability. DNase/RNase free and nonpyrogenic.
2,600.00 - ₹8,300.00INR / Case of 1000
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Contained Volume (µL) Overall Length (mm) Description
Availability Pricing
COMPARE
1 228 Lab Inoculating Needles, Sterile, Polystyrene, 1 µL, Bags of 10; 1000/Cs
₹8,300.00INR / Case of 1000
COMPARE
1 228 Lab Inoculating Needles, Sterile, Polystyrene, 1 µL, Individually Wrapped; 250/Cs
₹2,600.00INR / Case of 250
COMPARE
1 228 Lab Inoculating Needles, Sterile, Polystyrene, 1 µL, Bags of 25; 1000/Cs
₹6,900.00INR / Case of 1000