This product is now discontinued, View Related Products
Cole-Parmer® Digital Sanitary Pressure Gauges
Monitor pressure on food, beverage, and pharmaceutical lines
  • Certified to 3A sanitary standards
  • All-stainless wetted parts with flush diaphragm are easy to clean
  • NEMA 4X housing with coated circuit boards withstands washdown procedures
These digital sanitary pressure gauges feature a rugged ABS and polycarbonate housing with NEMA 4X rating for protection against strong jets of water. The circuitry is further protected against moisture by a conformal polymer coating, so these gauges are well-suited to beverage and brewery service. The fluid path is 316L stainless steel with a flush-mounted diaphragm and FDA-compliant fill solution to comply with 3A sanitary standards.
Call For Price
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Max Pressure (PSI) Power Source
Availability Pricing
COMPARE
15 Battery
Call For Price
COMPARE
30 Battery
Call For Price
COMPARE
60 Battery
Call For Price
COMPARE
100 Battery
Call For Price
COMPARE
15 Loop
Call For Price
COMPARE
30 Loop
Call For Price
COMPARE
60 Loop
Call For Price
COMPARE
100 Loop
Call For Price
Showing 1- 8 of 8