Cole-Parmer® WB-400 Series Digital General-Purpose Baths
Get digital accuracy at an analog price
  • Recall settings at the touch of a button—program up to 5 commonly used set points
  • Know when process is complete with built-in timer
  • Easy-to-read TFT display shows your choice of °C or °F
  • Features high-temperature alarm and safety shutdown
Cole-Parmer utility baths combine precise temperature control and excellent temperature stability to provide you with reliable heating. These baths feature dual thermostats — a primary microprocessor-based temperature controller that provides ±0.1°C stability (±0.2°C uniformity), and a secondary over-temperature safety sensor prevents samples from overheating. For optimal temperature accuracy, the primary controller can be calibrated to your reference thermometer.
The stainless steel tank is seamless—no welds to leak or rust—and the rounded corners provide an easy-to-clean interior. The high-dome, hinged polycarbonate lid tilts out of the way, while allowing complete view of the sample and condensation to drain back. The larger 10-, 20-, and 28-liter models also feature a rear-mounted drain.
Select operation in English, Spanish, French, Chinese, or German.
151,000.00 - ₹276,000.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Power (VAC) Reservoir Capacity (L)
Availability Pricing
COMPARE
100 120 2
₹151,000.00INR / Each
COMPARE
100 240 2
₹151,000.00INR / Each
COMPARE
100 120 5
₹182,000.00INR / Each
COMPARE
100 240 5
₹182,000.00INR / Each
COMPARE
100 120 10
₹217,000.00INR / Each
COMPARE
100 240 10
₹217,000.00INR / Each
COMPARE
100 120 20
₹262,000.00INR / Each
COMPARE
100 240 20
₹262,000.00INR / Each
COMPARE
100 120 28
₹276,000.00INR / Each
COMPARE
100 240 28
₹276,000.00INR / Each
Showing 1- 10 of 10