Cole-Parmer® CS-200 Series Plus Counting Scales
Quick and easy weighing in a compact, durable design
  • Three backlit LCDs show count, piece weight, and total weight
  • Sealed keypad protects against dirt and spills
  • Large stainless-steel pan allows for easy cleaning
  • Overload protection prevents damage to internal components
  • Programmable auto power-off saves energy
Affordable, compact, and portable, these scales offer fast parts counting wherever you need it. Take them through the warehouse or use them in your lab. Built-in rechargeable battery delivers up to 90 hours of operation on a single charge.
Scales are simple to operate with a numeric keypad for direct entry of piece weights. Weighing units include g, kg, lb, lb:oz, and oz. Other features include an internal memory, adjustable leveling feet, and low battery level indicator. The scales can also operate on a low voltage USB input.
47,200.00 - ₹72,000.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (kg) Readability (g)
Availability Pricing
COMPARE
External 3 0.1
₹47,200.00INR / Each
COMPARE
External 6 0.2
₹47,200.00INR / Each
COMPARE
External 15 0.5
₹47,200.00INR / Each
COMPARE
External 30 1
₹47,200.00INR / Each
COMPARE
External 3 0.1
₹47,200.00INR / Each
COMPARE
External 6 0.2
₹47,200.00INR / Each
COMPARE
External 15 0.5
₹72,000.00INR / Each
COMPARE
External 30 1
₹47,200.00INR / Each
Showing 1- 8 of 8