Cole-Parmer® Essentials Microscope Slide Cover Glasses
Protect your samples
  • Choose from two thicknesses
  • Constructed of borosilicate glass
Choose from two thicknesses: no. 1 (0.13 to 0.17 mm) or no.1.5 (0.16 to 0.19 mm). Cover glasses are made from borosilicate glasses and packaged by the ounce. The cover slips feature extra-white soda-lime glass with very low iron content.
3,000.00 - ₹3,100.00INR / Box of 156
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Size Thickness Description
Availability Pricing
COMPARE
Square (22 x 22 mm) No. 1 (0.13 to 0.17 mm) Cover glasses, 22 x 22 mm square, 0.13 to 0.17 mm thickness
₹3,000.00INR / Box of 156
COMPARE
Square (22 x 22 mm) No. 1.5 (0.16 to 0.19 mm) Cover glasses, 22 x 22 mm square, 0.16 to 0.19 mm thickness
₹3,100.00INR / Box of 138
COMPARE
Rectangular (22 x 40 mm) No. 1 (0.13 to 0.17 mm) Cover glasses, 22 x 40 mm rectangular, 0.13 to 0.17 mm thickness
₹3,000.00INR / Box of 87
COMPARE
Rectangular (22 x 40 mm) No. 1.5 (0.16 to 0.19 mm) Cover glasses, 22 x 40 mm rectangular, 0.16 to 0.19 mm thickness
₹3,100.00INR / Box of 76
COMPARE
Rectangular (24 x 60 mm) No. 1 (0.13 to 0.17 mm) Cover glasses, 24 x 60 mm rectangular, 0.13 to 0.17 mm thickness
₹3,000.00INR / Box of 54