This product is now discontinued, View Related Products
Cole-Parmer Cordless Stereo Microscopes
Compact size and cordless operation make these microscopes ideal for field use
  • Versatile top and bottom lights illuminate your sample
Compact microscopes adjust from 12" to 13-5/8" (30.5 to 34.6 cm) tall — they fit in tight spots and are easy to move where you need them. Cordless design allows field use and use in lab situations where an outlet isn't convenient. Dual-power microscopes feature parfocalled, parcentered, and achromatic objective lenses. Illumination control permits use of top and bottom lights together, top only, or bottom only.
Working distance is 75 mm and maximum specimen height is 35 mm. Stage is supplied with one transparent plate, one white plate, and one black plate for contrast viewing. Microscopes feature fixed arm, fixed head, and rugged cast-metal frame.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Head Configuration Magnification
Availability Pricing
COMPARE
Binocular 10x/30x
Call For Price
COMPARE
Binocular 20x/40x