This product is now discontinued, View Related Products
Cole-Parmer Cordless Compound Microscopes
Rechargeable NiMH batteries provide up to 50 hours of operation on an 8-hour charge
  • LED illumination with intensity control draws little power and provides heat-free lighting
Ease of use and precision optics make these microscopes ideal for the student and the professional. Binocular Siedentopf viewing head is inclined at 30° and rotates 360° for easy sharing. Objective turret is ball-bearing mounted for smooth magnification changes; reverse mounting allows easy access to specimens and stage.
Rechargeable NiMH batteries allow you to use this microscope in the field or where conventional power isn't convenient. Included recharger shuts off when batteries are fully charged, eliminating overcharging that can shorten battery life. Microscopes can be used during recharging, or recharger can be used as a power cord for 115 VAC operation.
Microscope features coarse and fine adjustments, widefield 10x eyepieces with pointer, 4-3/4" x 4-3/4" (121 x 121 mm) stage with specimen holder, and Abbe condenser with iris diaphragm.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Maximum Magnification Objectives
Availability Pricing
COMPARE
1000x Achromatic
Call For Price
COMPARE
1000x ASC