Cole-Parmer Compact Benchtop Ultraviolet Transilluminator
Compact design with large, 16 x 19 cm, viewing area
  • View gels at either 50% or 100%—dual intensity setting
  • 302 nm midrange UV operation—ideal for EtBr excitation
  • 8-watt UV bulb array provides even exposure across entire surface
  • Hinged cover opens 90° to accommodate documentation equipment
Optimize bench space with this UV transilluminator. The small footprint has an expanded exposure area for viewing larger gels, or multiple smaller ones. Generous black-glass filter surface is easy to clean, durable, and scratch resistant.
Dual intensity settings allow for observation at 50% or 100%—ideal for many applications. 302 nm wavelength output is compatible with ethidium bromide, Gel Red™, Gel Green™, SYBR® Green, SYBR® Safe, and Diamond™ Nucleic Acid Stains.
202,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (Hz) Power (VAC) Wavelength
Availability Pricing
COMPARE
50/60 115 302
₹202,000.00INR / Each
COMPARE
50/60 230 302
₹202,000.00INR / Each