This product is now discontinued, View Related Products
Cole-Parmer Combination Vertical Gel Systems
Increase throughput of your electrophoresis system – run up to four gels or four blots simultaneously in one unit!
  • Versatile unit accepts slab gels or Western blots
  • Designed for use with fast and efficient precast gels or traditional handcasting
  • Use with precast gels (see Related Product ad) to increase your throughput
These multipurpose gel systems are designed for use with either precast gels or traditional handcast gels to suit your needs. Sturdy design has no loose parts to keep track of and allows for easy loading and removal of your samples. Hinged-core design allows you to effortlessly extract the cassettes without removing the core from the tank—for quick sample processing and immediate reuse of buffers for your next set of samples. The color-coded anode and cathode safety features help with identification. Tank design includes a stir bar corral and hinged core.
Call For Price
21 variations of this product are available.
See All Product Options Below
21 PRODUCT OPTIONS
Compareitem specifications
Buffer Solution Required (mL) Gel Thickness (mm) Number of Wells
Availability Pricing
COMPARE
1000
Call For Price
COMPARE
1 10
Call For Price
COMPARE
1 14
Call For Price
COMPARE
1.5 10
Call For Price
COMPARE
1.5 14
Call For Price
COMPARE
0.75 10
Call For Price
COMPARE
0.75 14
Call For Price
COMPARE
1600
Call For Price
COMPARE
1600
Call For Price
COMPARE
1600 1 10
Call For Price
COMPARE
1600 1 14
Call For Price
COMPARE
1600 1.5 10
Call For Price
COMPARE
1600 1.5 14
Call For Price
COMPARE
1600 0.75 10
Call For Price
COMPARE
1600 0.75 14
Call For Price
COMPARE
1600 1 10
Call For Price
COMPARE
1600 1 14
Call For Price
COMPARE
1600 1.5 10
Call For Price
COMPARE
1600 1.5 14
Call For Price
COMPARE
1600 0.75 10
Call For Price
Showing 1 - 20 of 21