Cole-Parmer® Essentials Straight-Sided Round Jars, Clear Glass
Ideal for general lab use
  • Leakproof PTFE-lined caps protect contents
These straight-sided round bottles allow easy access and transfer of contents.
9,300.00 - ₹12,400.00INR / Case of 48
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Overall Height (in)
Availability Pricing
COMPARE
30 38-400 2 1/10
₹12,100.00INR / Case of 48
COMPARE
60 53-400 1 27/32
₹9,400.00INR / Case of 24
COMPARE
125 58-400 2 3/4
₹11,600.00INR / Case of 24
COMPARE
185 63-400 3 1/5
₹9,900.00INR / Case of 24
COMPARE
250 70-400 3 45/64
₹12,400.00INR / Case of 24
COMPARE
500 89-400 3 51/64
₹9,300.00INR / Case of 12
COMPARE
1000 89-400 6 51/64
₹9,900.00INR / Case of 12
Showing 1- 7 of 7