Cole-Parmer® Essentials Narrow-Mouth Bottles, Clear Glass
PTFE-lined bottle cap promotes bottle cleanliness
Economically priced glass bottles are priced to meet your budget requirements. Each bottle comes with an attached white foam backed, PTFE-lined bottle cap to promote bottle cleanliness.
6,100.00 - ₹10,800.00INR / Case of 48
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Outside Diameter (OD) (in)
Availability Pricing
COMPARE
30 20-400 1 1/4
₹10,800.00INR / Case of 48
COMPARE
60 20-400 1 1/2
₹6,100.00INR / Case of 24
COMPARE
125 22-400 1 7/8
₹7,200.00INR / Case of 24
COMPARE
250 24-400 2 3/8
₹9,200.00INR / Case of 24
COMPARE
500 28-400 3
₹6,100.00INR / Case of 12
COMPARE
1000 33-400 3 3/4
₹10,200.00INR / Case of 12